• USA Home
  • D6154 - 4,15-Diacetylverrucarol

EMAIL THIS PAGE TO A FRIEND
D6154 Sigma

4,15-Diacetylverrucarol

Synonym: 4β,15-Diacetoxy-12,13-epoxytrichothec-9-ene, Compound A-2

  • CAS Number 2198-94-9

  • Empirical Formula (Hill Notation) C19H26O6

  • Molecular Weight 350.41

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?