EMAIL THIS PAGE TO A FRIEND
D6549 Sigma-Aldrich

Sulfamoxole

≥80%

Synonym: 4-Amino-N-(4,5-dimethyl-2-oxazolyl)benzenesulfonamide, N1-(4,5-Dimethyloxazol-2-yl)sulfanilamide

  • CAS Number 729-99-7

  • Empirical Formula (Hill Notation) C11H13N3O3S

  • Molecular Weight 267.30

  •  Beilstein/REAXYS Number 248434

  •  EC Number 211-982-7

  •  MDL number MFCD00005303

Purchase

Properties

assay   ≥80%
SMILES string   Cc1nc(NS(=O)(=O)c2ccc(N)cc2)oc1C
InChI   1S/C11H13N3O3S/c1-7-8(2)17-11(13-7)14-18(15,16)10-5-3-9(12)4-6-10/h3-6H,12H2,1-2H3,(H,13,14)
InChI key   CYFLXLSBHQBMFT-UHFFFAOYSA-N
Safety & Documentation

Safety Information

WGK Germany 
2
RTECS 
WO9150000
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?