• USA Home
  • D6784 - 2-Deoxy-D-Glucose-1-14C

EMAIL THIS PAGE TO A FRIEND
D6784 Sigma-Aldrich

2-Deoxy-D-Glucose-1-14C

suitable for brain imaging studies, ≥98% radiochemical purity basis (HPLC), aqueous solution

Purchase

Properties

assay   ≥98% radiochemical purity basis (HPLC)
form   aqueous solution
extent of labeling   0.5 mCi per mL
  40-60 mCi per mmol
suitability   suitable for brain imaging studies
mass shift   M+2
shipped in   dry ice
storage temp.   −20°C

Description

Packaging

Combi-vial

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?