• USA Home
  • D6876 - N,9-O-Diacetylneuraminic acid ammonium salt

EMAIL THIS PAGE TO A FRIEND
D6876 Sigma-Aldrich

N,9-O-Diacetylneuraminic acid ammonium salt

~95% (HPLC)

Synonym: 9-O-Acetylsialic acid

  • CAS Number 148067-22-5

  • Empirical Formula (Hill Notation) C13H21NO10

  • Molecular Weight 351.31

Purchase

Properties

assay   ~95% (HPLC)
storage temp.   −20°C

Description

Biochem/physiol Actions

Determinant of cell surface receptors for influenza viruses.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?