• USA Home
  • D6912 - (±)-Deoxyephedrine hydrochloride

EMAIL THIS PAGE TO A FRIEND
D6912 Sigma

(±)-Deoxyephedrine hydrochloride

Synonym: (±)-Methamphetamine, (±)-2-(Methylamino)-1-phenylpropane

  • Empirical Formula (Hill Notation) C10H15N · HCl

  • Molecular Weight 185.69

  •  EC Number 206-093-6

  •  MDL number MFCD00069952

Purchase

Properties

form   neat
drug control   USDEA Schedule II; Home Office Schedule 2; stupéfiant; kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Biochem/physiol Actions

Sympathomimetic; more potent central effects than amphetamine.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
SH5075000

Documents

Certificate of Analysis

Protocols & Articles

Articles

HPLC Analysis Report of Amphetamine and Methamphetamine Enantiomers on Astec® CHIROBIOTIC® V2

From our library of Articles, Sigma-Aldrich presents HPLC Analysis Report of Amphetamine and Methamphetamine Enantiomers on Astec® CHIROBIOTIC® V2
Keywords: Chromatography, Clinical, Forensic, High performance liquid chromatography, Pharmaceutical

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?