• USA Home
  • D6915 - 13,14-Dihydro-15-ketoprostaglandin E2 solution

EMAIL THIS PAGE TO A FRIEND
D6915 Sigma

13,14-Dihydro-15-ketoprostaglandin E2 solution

≥95%, 10 mg/mL in methyl acetate

  • CAS Number 363-23-5

  • Empirical Formula (Hill Notation) C20H32O5

  • Molecular Weight 352.47

  •  MDL number MFCD00135217

Purchase

Properties

InChI Key   CUJMXIQZWPZMNQ-XYYGWQPLSA-N
assay   ≥95%
concentration   10 mg/mL in methyl acetate
shipped in   dry ice
storage temp.   −20°C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1231 - class 3 - PG 2 - Methyl acetate
WGK Germany 
2
Flash Point(F) 
8.6 °F
Flash Point(C) 
-13 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?