EMAIL THIS PAGE TO A FRIEND
D7020 Sigma-Aldrich

Dihydrotachysterol

Synonym: 9,10-Secoergosta-5,7,22-trien-3β-ol, DHT2

  • CAS Number 67-96-9

  • Empirical Formula (Hill Notation) C28H46O

  • Molecular Weight 398.66

  •  EC Number 200-672-7

Purchase

Properties

form   solid
storage temp.   −20°C
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
WW0600000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?