EMAIL THIS PAGE TO A FRIEND
D7138 Sigma

DL-Dihydrozeatin

~90%

  • CAS Number 23599-75-9

  • Empirical Formula (Hill Notation) C10H15N5O

  • Molecular Weight 221.26

  •  Beilstein Registry Number 1215977

  •  MDL number MFCD00056912

Purchase

Properties

InChI Key   XXFACTAYGKKOQB-UHFFFAOYSA-N
assay   ~90%
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?