• USA Home
  • D7177 - Difucosyl-para-lacto-N-hexaose from human milk

EMAIL THIS PAGE TO A FRIEND
D7177 Sigma-Aldrich

Difucosyl-para-lacto-N-hexaose from human milk

≥85% (HPLC)

Synonym: DFpLNH, Lea-Lex-Lac

  • CAS Number 64309-01-9

  • Empirical Formula (Hill Notation) C52H88N2O39

  • Molecular Weight 1365.25

  •  MDL number MFCD01076421

  •  PubChem Substance ID 24894088

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates D-F, Oligosaccharide More...
biological source   human milk
assay   ≥85% (HPLC)
form   powder
storage temp.   2-8°C
SMILES string   CC1OC(OC2C(CO)OC(OC3C(O)C(CO)OC(OC4C(CO)OC(OC5C(O)C(CO)OC(OC(C(O)CO)C(O)C(O)C=O)C5O)C(NC(C)=O)C4OC6OC(C)C(O)C(O)C6O)C3O)C(NC(C)=O)C2OC7OC(CO)C(O)C(O)C7O)C(O)C(O)C1O
InChI   1S/C52H88N2O39/c1-12-25(66)31(72)34(75)48(80-12)88-40-21(10-60)85-47(24(54-15(4)63)43(40)91-50-36(77)33(74)28(69)18(7-57)82-50)93-45-30(71)20(9-59)84-52(38(45)79)89-41-22(11-61)86-46(23(53-14(3)62)42(41)90-49-35(76)32(73)26(67)13(2)81-49)92-44-29(70)19(8-58)83-51(37(44)78)87-39(17(65)6-56)27(68)16(64)5-55/h5,12-13,16-52,56-61,64-79H,6-11H2,1-4H3,(H,53,62)(H,54,63)
InChI key   BCUMESVDMXHZRL-UHFFFAOYSA-N
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?