EMAIL THIS PAGE TO A FRIEND
D7634 Sigma-Aldrich

Dansylhydrazine

≥90%

Synonym: 5-(Dimethylamino)-1-naphthalenesulfonic hydrazide, 5-(Dimethylamino)naphthalene-1-sulfono­hydra­zide, Dns-Hz

  • CAS Number 33008-06-9

  • Linear Formula (CH3)2NC10H6SO2NHNH2

  • Molecular Weight 265.33

  •  Beilstein/REAXYS Number 2868662

  •  EC Number 251-337-7

  •  MDL number MFCD00003986

  •  PubChem Substance ID 24894145

Purchase

Properties

assay   ≥90%
mp   126-130 °C (lit.)
storage temp.   −20°C
SMILES string   CN(C)c1cccc2c(cccc12)S(=O)(=O)NN
InChI   1S/C12H15N3O2S/c1-15(2)11-7-3-6-10-9(11)5-4-8-12(10)18(16,17)14-13/h3-8,14H,13H2,1-2H3
InChI key   KPQYDVAFRDWIBW-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

30434 Dansylhydrazine, BioReagent, suitable for fluorescence, ≥90% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?