• USA Home
  • D8003 - 6,6′-Dithiodi(2-naphthol)

EMAIL THIS PAGE TO A FRIEND
D8003 Sigma-Aldrich

6,6′-Dithiodi(2-naphthol)

Synonym: 6,6′-Dihydroxy-2,2′-dinaphthyl disulfide, 6-Hydroxy-2-naphthyl disulfide, Bis(6-hydroxy-2-naphthyl) disulfide, DDD

  • CAS Number 6088-51-3

  • Empirical Formula (Hill Notation) C20H14O2S2

  • Molecular Weight 350.45

  •  Beilstein/REAXYS Number 3398968

  •  EC Number 228-025-4

  •  MDL number MFCD00004083

Purchase

Properties

storage temp.   −20°C

Description

Application

Useful for the determination of sulfhydryl groups in proteins.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?