EMAIL THIS PAGE TO A FRIEND
D8400 Sigma

Dibromomethane-d2

Synonym: Methylene-d2 bromide

Purchase

Properties

vapor density   5.9 (vs air)
InChI Key   FJBFPHVGVWTDIP-DICFDUPASA-N
isotopic purity   99 atom % D
mol wt   mol wt 175.83 by atom % calculation
refractive index   n20/D 1.5381(lit.)
bp   99 °C(lit.)
mass shift   M+2
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2664 6.1 / PGIII
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?