• USA Home
  • D8639 - 1,2-Dipalmitoyl-3-iodo-3-deoxy-rac-glycerol

EMAIL THIS PAGE TO A FRIEND
D8639 Sigma-Aldrich

1,2-Dipalmitoyl-3-iodo-3-deoxy-rac-glycerol

Purchase

Properties

Related Categories Biochemicals and Reagents, Diacylglycerols, Glycerolipids, Lipids
InChI Key   JJKDQFHYOKPMCL-UHFFFAOYSA-N
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?