• USA Home
  • D8663 - 1,7-Dimethyluric acid

EMAIL THIS PAGE TO A FRIEND
D8663 Sigma-Aldrich

1,7-Dimethyluric acid

~95% (HPLC)

Synonym: 1,7-Dimethyl-2,6,8-trihydroxypurine

  • CAS Number 33868-03-0

  • Empirical Formula (Hill Notation) C7H8N4O3

  • Molecular Weight 196.16

  •  Beilstein/REAXYS Number 219682

  •  EC Number 251-706-2

  •  MDL number MFCD00005713

  •  PubChem Substance ID 24894111

Purchase

Properties

InChI Key   NOFNCLGCUJJPKU-UHFFFAOYSA-N
assay   ~95% (HPLC)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?