• USA Home
  • D8769 - 1,2-Dimyristoyl-rac-glycerol

EMAIL THIS PAGE TO A FRIEND
D8769 Sigma-Aldrich

1,2-Dimyristoyl-rac-glycerol

~99%

Synonym: 1,2-Dimyristin, 1,2-Ditetradecanoyl-rac-glycerol

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Diacylglycerols, Glycerolipid Metabolism, Glycerolipids,
assay   ~99%
impurities   ~1% 1,3-isomer
storage temp.   −20°C
SMILES string   CCCCCCCCCCCCCC(=O)OCC(CO)OC(=O)CCCCCCCCCCCCC
InChI   1S/C15H28O5/c1-3-5-7-9-14(17)19-12-13(11-16)20-15(18)10-8-6-4-2/h13,16H,3-12H2,1-2H3
InChI key   DRUFTGMQJWWIOL-UHFFFAOYSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?