EMAIL THIS PAGE TO A FRIEND
D8781 Sigma-Aldrich

Dmt-thymidine

Synonym: 5′-O-(4,4′-Dimethoxytrityl)thymidine, DMT-T

  • CAS Number 40615-39-2

  • Empirical Formula (Hill Notation) C31H32N2O7

  • Molecular Weight 544.59

  •  Beilstein/REAXYS Number 599297

  •  MDL number MFCD00010113

Purchase

Properties

mp   114-116 °C (subl.) (lit.)
storage temp.   −20°C
SMILES string   COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2O)N3C=C(C)C(=O)NC3=O)(c4ccccc4)c5ccc(OC)cc5
InChI   1S/C31H32N2O7/c1-20-18-33(30(36)32-29(20)35)28-17-26(34)27(40-28)19-39-31(21-7-5-4-6-8-21,22-9-13-24(37-2)14-10-22)23-11-15-25(38-3)16-12-23/h4-16,18,26-28,34H,17,19H2,1-3H3,(H,32,35,36)/t26-,27+,28+/m0/s1
InChI key   UBTJZUKVKGZHAD-UPRLRBBYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?