EMAIL THIS PAGE TO A FRIEND
D8793 Sigma-Aldrich

1,2-Dichloroethane

ACS reagent

Synonym: Ethylene chloride, Ethylene dichloride

  • CAS Number 107-06-2

  • Linear Formula ClCH2CH2Cl

  • Molecular Weight 98.96

  •  Beilstein/REAXYS Number 605264

  •  EC Number 203-458-1

  •  MDL number MFCD00000963

Purchase

Properties

grade   ACS reagent
vapor density   3.4 (20 °C, vs air)
vapor pressure   87 mmHg ( 25 °C)
InChI Key   WSLDOOZREJYCGB-UHFFFAOYSA-N
autoignition temp.   775 °F
expl. lim.   15.6 %
impurities   ≤0.0003 meq/g Titratable acid
  ≤0.03% Water (H2O)
evapn. residue   ≤0.002%
color   APHA: ≤10
refractive index   n20/D 1.444 (lit.)
bp   82.5-84.5 °C (lit.)
  83 °C (lit.)
mp   −35 °C (lit.)
density   1.256 g/mL at 25 °C (lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
UN 1184 3/PG 2
WGK Germany 
3
RTECS 
KI0525000
Flash Point(F) 
55 °F
Flash Point(C) 
13 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?