• USA Home
  • D9148 - [D-Arg6]-Dynorphin A Porcine fragment 1-13

EMAIL THIS PAGE TO A FRIEND
D9148 Sigma-Aldrich

[D-Arg6]-Dynorphin A Porcine fragment 1-13

≥97% (HPLC)

  • CAS Number 75921-87-8

  • Empirical Formula (Hill Notation) C75H126N24O15

  • Molecular Weight 1603.95

  •  MDL number MFCD00076369

  •  PubChem Substance ID 24894260

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Dynorphins, Opioid Peptides, Peptides and Proteins,
InChI Key   OVVIBUHLQIYUEU-UHFFFAOYSA-N
assay   ≥97% (HPLC)
composition   Peptide content, ~70%
storage temp.   −20°C
Gene Information   pig ... PDYN(445529)

Description

Amino Acid Sequence

Tyr-Gly-Gly-Phe-Leu-D-Arg-Arg-Ile-Arg-Pro-Lys-Leu-Lys

Biochem/physiol Actions

κ opioid receptor agonist; anticonvulsant

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?