• USA Home
  • D9192 - 4,5-Dimethoxy-6-nitrobenzaldehyde

EMAIL THIS PAGE TO A FRIEND
D9192 Sigma

4,5-Dimethoxy-6-nitrobenzaldehyde

≥98% (HPLC), solid

Synonym: 3,4-Dimethoxy-6-nitrobenzaldehyde, 6-Nitroveratraldehyde, DMNB

Purchase

Properties

assay   ≥98% (HPLC)
form   solid
color   yellow
mp   133.2-134.5 °C
solubility   DMSO: 22 mg/mL
storage temp.   2-8°C

Description

Biochem/physiol Actions

DNA-dependent protein kinase (DNA-PK) inhibitor (IC50 = 15 μM), an enzyme involved in the non-homologous end-joining (NHEJ) pathway of double-stranded DNA break (DSB) repair in human cells.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?