• USA Home
  • D9254 - DNP-L-glutamic acid bis(cyclohexylammonium) salt

EMAIL THIS PAGE TO A FRIEND
D9254 Sigma-Aldrich

DNP-L-glutamic acid bis(cyclohexylammonium) salt

Synonym: N-(2,4-Dinitrophenyl)-L-glutamic acid bis(cyclohexylammonium) salt

  • CAS Number 102783-75-5

  • Empirical Formula (Hill Notation) C11H11N3O8 · 2C6H13N

  • Molecular Weight 511.57

  •  PubChem Substance ID 329798897

Purchase

Properties

Related Categories Amino Acid Analysis, Amino Acid Analysis Reagents, Biochemicals and Reagents, Peptide Analysis and Characterization, Peptides More...
InChI Key   DCNBQPUKOWUYTM-JZGIKJSDSA-N
storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?