• USA Home
  • E1758 - β-Estradiol 17-propionate

EMAIL THIS PAGE TO A FRIEND
E1758 Sigma-Aldrich

β-Estradiol 17-propionate

≥98%

Synonym: 1,3,5(10)-Estratriene-3,17β-diol 17-propionate, 3,17β-Dihydroxy-1,3,5(10)-estratriene 17-propionate

  • CAS Number 3758-34-7

  • Empirical Formula (Hill Notation) C21H28O3

  • Molecular Weight 328.45

  •  Beilstein/REAXYS Number 3215697

  •  EC Number 223-164-7

  •  MDL number MFCD00056539

Purchase

Properties

assay   ≥98%
SMILES string   CCC(=O)OC1CCC2C3CCc4cc(O)ccc4C3CCC12C
InChI   1S/C21H28O3/c1-3-20(23)24-19-9-8-18-17-6-4-13-12-14(22)5-7-15(13)16(17)10-11-21(18,19)2/h5,7,12,16-19,22H,3-4,6,8-11H2,1-2H3
InChI key   PQCRZWCSVWBYSC-UHFFFAOYSA-N
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?