• USA Home
  • E2145 - Express-Ion exchanger D free base

EMAIL THIS PAGE TO A FRIEND
E2145 Sigma-Aldrich

Express-Ion exchanger D free base

60-130 μm

Synonym: Cellulose DEAE, DEAE-Cellulose, Diethylaminoethyl-cellulose

Purchase

Properties

Related Categories Analytical/Chromatography, Cation Exchange Media, Ion Exchange Chromatography, LPLC/TLC/Paper Chromatography, Molecular Biology,
InChI Key   GUBGYTABKSRVRQ-WFVLMXAXSA-N
feature   flow rate 150 cm/hr
matrix active group   DEAE (diethylaminoethyl)
particle size   60-130 μm
pH range   2 - 9
capacity   1 meq/g

Description

Application

Express-Ion exchangers are weakly basic anion exchangers used in protein chromatography, ion exchange chromatography, cation exchange media, cellulose ion exchange TLC adsorbents and DEAE-cellulose anion exchange TLC adsorbents.

Legal Information

Express-Ion is a trademark of Whatman International Ltd.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?