• USA Home
  • E3880 - Ethyl acetimidate hydrochloride

EMAIL THIS PAGE TO A FRIEND
E3880 Sigma-Aldrich

Ethyl acetimidate hydrochloride

  • CAS Number 2208-07-3

  • Linear Formula CH3C(=NH)OC2H5 · HCl

  • Molecular Weight 123.58

  •  Beilstein/REAXYS Number 3552401

  •  EC Number 218-631-7

  •  MDL number MFCD00012572

Purchase

Properties

mp   112-114 °C (lit.)
storage temp.   2-8°C
SMILES string   Cl.CCOC(C)=N
InChI   1S/C4H9NO.ClH/c1-3-6-4(2)5;/h5H,3H2,1-2H3;1H
InChI key   WGMHMVLZFAJNOT-UHFFFAOYSA-N
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

188840 Ethyl acetimidate hydrochloride, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?