EMAIL THIS PAGE TO A FRIEND
E5402 Sigma-Aldrich

Etbicythionat

≥98% (NMR), solid

Synonym: 2,2-Bis(hydroxymethyl)butanol O,O′,O″-phosphorothioate

  • CAS Number 224790-71-0

  • Empirical Formula (Hill Notation) C6H11O3PS

  • Molecular Weight 194.19

  •  MDL number MFCD00236474

Purchase

Properties

assay   ≥98% (NMR)
form   solid
SMILES string   CC[C@@]12COP(=S)(OC1)OC2

Description

Biochem/physiol Actions

A potent non-competitive GABA antagonist in mammals; causes epileptiform seizures.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?