• USA Home
  • E5506 - Methyl all-cis-5,8,11,14,17-eicosapentaenoate

EMAIL THIS PAGE TO A FRIEND
E5506 Sigma-Aldrich

Methyl all-cis-5,8,11,14,17-eicosapentaenoate

90-95%, liquid

Synonym: cis-5,8,11,14,17-Eicosapentaenoic acid methyl ester

  • CAS Number 2734-47-6

  • Linear Formula CH3(CH2CH=CH)5(CH2)3CO2CH3

  • Molecular Weight 316.48

  •  Beilstein Registry Number 1914828

  •  MDL number MFCD02259226

Purchase

Properties

InChI Key   QWDCYFDDFPWISL-JEBPEJKESA-N
assay   90-95%
form   liquid
storage temp.   −20°C

Description

Packaging

Sealed ampule.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
219.2 °F
Flash Point(C) 
104 °C
Protocols & Articles

Articles

Omega-3 Fatty Acids and Heart Disease

The potential for the prevention and treatment of cardiovascular disease through increased dietary intake of omega-3 (w-3) fish oils is not a recent scientific discovery. A historically important stu...
BioFiles 2007, 2.2, 14.
Keywords: Anti-inflammatory agents, Biochemistry, Cardiovascular, Clinical, Degradations, Esterifications, Immunology, Inflammation, Lipid Metabolism, Metabolism, Metabolites, Methods, Nutrition, Oxidations, Transcription

Related Products

replaced by

Product #

Description

Add to Cart

E2012 Methyl all-cis-5,8,11,14,17-eicosapentaenoate, ≥97% (capillary GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?