EMAIL THIS PAGE TO A FRIEND
E7132 Sigma-Aldrich

Estrone acetate

~95%

Synonym: 1,3,5(10)-Estratrien-17-one 3-acetate, 3-Acetoxy-1,3,5(10)-estratrien-17-one

  • CAS Number 901-93-9

  • Empirical Formula (Hill Notation) C20H24O3

  • Molecular Weight 312.40

  •  MDL number MFCD00056411

Purchase

Properties

assay   ~95%
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?