• USA Home
  • E7876 - Epiandrosterone sulfate sodium salt

EMAIL THIS PAGE TO A FRIEND
E7876 Sigma-Aldrich

Epiandrosterone sulfate sodium salt

Synonym: 3β-Hydroxy-5α-androstan-17-one 3-sulfate, 5α-Androstan-3β-ol-17-one sulfate, Isoandrosterone sulfate

  • CAS Number 22229-22-7

  • Linear Formula C19H29O5SNa

  • Molecular Weight 392.49

Purchase

Properties

drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?