• USA Home
  • E8000 - Etiocholan-3α-ol-17-one glucuronide

EMAIL THIS PAGE TO A FRIEND
E8000 Sigma-Aldrich

Etiocholan-3α-ol-17-one glucuronide

Synonym: 3α-Hydroxy-5β-androstan-17-one 3-glucuronide, 5β-Androstan-3α-ol-17-one 3-glucuronide

  • CAS Number 3602-09-3

  • Empirical Formula (Hill Notation) C25H38O8

  • Molecular Weight 466.56

  •  MDL number MFCD00067126

  •  PubChem Substance ID 24894662

Purchase

Properties

Related Categories Androgen, Cell Biology, Cell Signaling and Neuroscience, Cytokines, Growth Factors and Hormones, Hormones,
form   solid
drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
shipped in   ambient
storage temp.   −20°C
SMILES string   CC12CCC(CC1CCC3C2CCC4(C)C3CCC4=O)OC5OC(C(O)C(O)C5O)C(O)=O
InChI   1S/C25H38O8/c1-24-9-7-13(32-23-20(29)18(27)19(28)21(33-23)22(30)31)11-12(24)3-4-14-15-5-6-17(26)25(15,2)10-8-16(14)24/h12-16,18-21,23,27-29H,3-11H2,1-2H3,(H,30,31)
InChI key   VFUIRAVTUVCQTF-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?