• USA Home
  • E8252 - 1,N6-Ethenoadenosine hydrochloride

EMAIL THIS PAGE TO A FRIEND
E8252 Sigma-Aldrich

1,N6-Ethenoadenosine hydrochloride

≥98% (HPLC)

  • CAS Number 36207-54-2

  • Empirical Formula (Hill Notation) C12H13N5O4 · HCl

  • Molecular Weight 327.72

  •  MDL number MFCD00038986

Purchase

Properties

assay   ≥98% (HPLC)
storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?