• USA Home
  • E9384 - [Cys(Acm)20,31]-Epidermal Growth Factor Fragment 20-31

EMAIL THIS PAGE TO A FRIEND
E9384 Sigma-Aldrich

[Cys(Acm)20,31]-Epidermal Growth Factor Fragment 20-31

≥97% (HPLC)

Synonym: Cys(S-Acetamidomethyl)-Met-His-Ile-Glu-Ser-Leu-Asp-Ser-Tyr-Thr-Cys(Acm)

  • CAS Number 89991-90-2

  • Empirical Formula (Hill Notation) C63H98N16O23S3

  • Molecular Weight 1543.74

  •  MDL number MFCD00133946

  •  PubChem Substance ID 24894695

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
InChI Key   UEJSENUVDZJGAP-UHFFFAOYSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C
Gene Information   human ... EGF(1950)

Description

Biochem/physiol Actions

The fragment of mouse epidermal growth factor (EGF) that contains the receptor-binding region as well as the predominant EGF antigenic determinant. This fragment exhibits the biological activity of EGF in vitro.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?