• USA Home
  • E9625 - β-Estriol 3-methyl ether

EMAIL THIS PAGE TO A FRIEND
E9625 Sigma-Aldrich

β-Estriol 3-methyl ether

~90%

Synonym: 1,3,5(10)-Estratriene-3,16α,17β-triol 3-methyl ether, 3,16α,17β-Trihydroxy-1,3,5(10)-estratriene 3-methyl ether

  • CAS Number 1474-53-9

  • Empirical Formula (Hill Notation) C19H26O3

  • Molecular Weight 302.41

  •  MDL number MFCD00046284

Purchase

Properties

assay   ~90%
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
KG7540000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?