• USA Home
  • E9626 - (−)-Pseudoephedrine

EMAIL THIS PAGE TO A FRIEND
E9626 Supelco

(−)-Pseudoephedrine

Synonym: (−)-ψ-Ephedrine, (1R,2R)-2-Methylamino-1-phenyl-1-propanol, L-Pseudoephedrine

  • CAS Number 321-97-1

  • Empirical Formula (Hill Notation) C10H15NO

  • Molecular Weight 165.23

  •  Beilstein/REAXYS Number 2803417

  •  EC Number 206-292-8

  •  MDL number MFCD00002203

Purchase

Properties

form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
SMILES string   CN[C@H](C)[C@H](O)c1ccccc1
InChI   1S/C10H15NO/c1-8(11-2)10(12)9-6-4-3-5-7-9/h3-8,10-12H,1-2H3/t8-,10+/m1/s1
InChI key   KWGRBVOPPLSCSI-SCZZXKLOSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Less active isomer of ψ-ephedrine

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
UL5750500

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Articles

HPLC Analysis of Pseudoephedrine Enantiomers on Astec® CHIROBIOTIC® T

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Pseudoephedrine Enantiomers on Astec® CHIROBIOTIC® T
Keywords: Chromatography, High performance liquid chromatography, Pharmaceutical

Related Products

replaced by

Product #

Description

Add to Cart

287644 (1R,2R)-(−)-Pseudoephedrine, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?