EMAIL THIS PAGE TO A FRIEND
F0134 Sigma-Aldrich

Fmoc-Ile-OH

Synonym: N-(9-Fluorenylmethoxycarbonyl)-L-isoleucine, Fmoc-L-isoleucine

  • CAS Number 71989-23-6

  • Empirical Formula (Hill Notation) C21H23NO4

  • Molecular Weight 353.41

  •  Beilstein/REAXYS Number 4716717

  •  EC Number 276-255-9

  •  MDL number MFCD00037125

Purchase

Properties

mp   145-147 °C (lit.)
storage temp.   −20°C
SMILES string   CC[C@H](C)[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O
InChI   1S/C21H23NO4/c1-3-13(2)19(20(23)24)22-21(25)26-12-18-16-10-6-4-8-14(16)15-9-5-7-11-17(15)18/h4-11,13,18-19H,3,12H2,1-2H3,(H,22,25)(H,23,24)/t13-,19-/m0/s1
InChI key   QXVFEIPAZSXRGM-DJJJIMSYSA-N
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

47628 Fmoc-Ile-OH, ≥98.0% (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?