• USA Home
  • F1012 - N-Formyl-Met-Leu-Phe-p-fluorobenzylamide

EMAIL THIS PAGE TO A FRIEND
F1012 Sigma-Aldrich

N-Formyl-Met-Leu-Phe-p-fluorobenzylamide

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Chemotactic Peptides, Immunomodulators, Peptides and Proteins,
storage temp.   −20°C
SMILES string   CSCCC(NC=O)C(=O)NC(CC(C)C)C(=O)NC(Cc1ccccc1)C(=O)NCc2ccc(F)cc2
InChI   1S/C28H37FN4O4S/c1-19(2)15-24(32-27(36)23(31-18-34)13-14-38-3)28(37)33-25(16-20-7-5-4-6-8-20)26(35)30-17-21-9-11-22(29)12-10-21/h4-12,18-19,23-25H,13-17H2,1-3H3,(H,30,35)(H,31,34)(H,32,36)(H,33,37)
InChI key   ABCCPEXHAJFNOT-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?