EMAIL THIS PAGE TO A FRIEND
F1383 Sigma-Aldrich

Fmoc-Ser(tBu)-OH

Synonym: Fmoc-O-tert-butyl-L-serine

  • CAS Number 71989-33-8

  • Empirical Formula (Hill Notation) C22H25NO5

  • Molecular Weight 383.44

  •  Beilstein/REAXYS Number 3632013

  •  EC Number 276-260-6

  •  MDL number MFCD00037127

Purchase

Properties

InChI Key   REITVGIIZHFVGU-IBGZPJMESA-N
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

47619 Fmoc-Ser(tBu)-OH, ≥98.0% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?