EMAIL THIS PAGE TO A FRIEND
F1393 Sigma-Aldrich

Fmoc-Asn(Trt)-OH

~95%

Synonym: Nα-(9-Fluorenylmethoxycarbonyl)-Nγ-trityl-L-asparagine, Nα-Fmoc-Nγ-trityl-L-asparagine

  • CAS Number 132388-59-1

  • Empirical Formula (Hill Notation) C38H32N2O5

  • Molecular Weight 596.67

  •  Beilstein Registry Number 4343823

  •  MDL number MFCD00077049

  •  PubChem Substance ID 329799678

Purchase

Properties

InChI Key   KJYAFJQCGPUXJY-UMSFTDKQSA-N
assay   ~95%
mp   201-204 °C (lit.)
storage temp.   −20°C
Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Hazard statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

47672 Fmoc-Asn(Trt)-OH, ≥97.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?