EMAIL THIS PAGE TO A FRIEND
F1501 Sigma-Aldrich

6-Fluorogramine

Synonym: 6-Fluoro-3-(dimethylaminomethyl)indole

  • CAS Number 343-93-1

  • Empirical Formula (Hill Notation) C11H13FN2

  • Molecular Weight 192.23

  •  MDL number MFCD00056918

  •  PubChem Substance ID 24894769

Purchase

Properties

Related Categories Biochemicals and Reagents, Luminescent Compounds/Detection, Other luminescent compounds More...
storage temp.   2-8°C
SMILES string   CN(C)Cc1c[nH]c2cc(F)ccc12
InChI   1S/C11H13FN2/c1-14(2)7-8-6-13-11-5-9(12)3-4-10(8)11/h3-6,13H,7H2,1-2H3
InChI key   PAAOUYLDLVHKKR-UHFFFAOYSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?