EMAIL THIS PAGE TO A FRIEND
F1883 Sigma-Aldrich

Fmoc-Gln-OH

Synonym: Fmoc-L-glutamine

  • CAS Number 71989-20-3

  • Empirical Formula (Hill Notation) C20H20N2O5

  • Molecular Weight 368.38

  •  Beilstein/REAXYS Number 4722773

  •  EC Number 276-254-3

  •  MDL number MFCD00037137

Purchase

Properties

InChI Key   IZKGGDFLLNVXNZ-KRWDZBQOSA-N
mp   220 °C (dec.) (lit.)
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?