• USA Home
  • F3387 - N-Formyl-DL-phenylalanine

EMAIL THIS PAGE TO A FRIEND
F3387 Sigma-Aldrich

N-Formyl-DL-phenylalanine

  • CAS Number 4289-95-6

  • Empirical Formula (Hill Notation) C10H11NO3

  • Molecular Weight 193.20

  •  MDL number MFCD00063281

Purchase

Properties

storage temp.   −20°C
SMILES string   OC(=O)C(Cc1ccccc1)NC=O
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?