• USA Home
  • F3504 - 5-Fluorocytosine arabinoside

EMAIL THIS PAGE TO A FRIEND
F3504 Sigma

5-Fluorocytosine arabinoside

  • CAS Number 4298-10-6

  • Empirical Formula (Hill Notation) C9H12FN3O5

  • Molecular Weight 261.21

  •  MDL number MFCD00036765

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
HA5450000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?