• USA Home
  • F3635 - Fibroblast Growth Factor, Acidic, Fragment 1-11 bovine

EMAIL THIS PAGE TO A FRIEND
F3635 Sigma-Aldrich

Fibroblast Growth Factor, Acidic, Fragment 1-11 bovine

≥97% (HPLC)

  • CAS Number 62031-54-3

  • Empirical Formula (Hill Notation) C62H95N15O15

  • Molecular Weight 1290.51

  •  MDL number MFCD00076545

Purchase

Properties

assay   ≥97% (HPLC)
storage temp.   −20°C
SMILES string   CC(C)C[C@H](NC(=O)[C@@H]1CCCN1C(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(N)=O)NC(=O)[C@@H](N)Cc2ccccc2)C(=O)NCC(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](Cc3ccc(O)cc3)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)N4CCC[C@H]4C(O)=O
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
RTECS 
LK3641300
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?