EMAIL THIS PAGE TO A FRIEND
F3766 Sigma

Firefly diluent

lyophilized powder

Purchase

Description

Application

For use in ATP determinations

Other Notes

Glycine diluent with added luciferin

Reconstitution

When reconstituted with 10 ml distilled water, vial will contain 50 mM glycine, 0.15 mM luciferin, 1 mM Tris, pH 7.6, 5 mM MgSO4, 0.55 mM EDTA, 0.1% (w/v) BSA, 0.1% (W/V) sodium azide

Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles

Protocols

Enzymatic Assay and ATP Sensitivity Test of Luciferase

To standardize a procedure to determine the enzymatic activity of Luciferase and/or to determine the ATP detection limit of Luciferase.
Keywords: Biochemistry, Enzyme activity, Enzymology, Size-exclusion chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?