• USA Home
  • F3792 - N-(3-[2-Furyl]acryloyl)-S-farnesyl-L-cysteine

EMAIL THIS PAGE TO A FRIEND
F3792 Sigma-Aldrich

N-(3-[2-Furyl]acryloyl)-S-farnesyl-L-cysteine

≥90% (TLC)

Purchase

Properties

Related Categories Activity, Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Fluorogenic More...
InChI Key   YEWYGZOGAGEKLT-KEBCOVAKSA-N
assay   ≥90% (TLC)
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?