• USA Home
  • F5895 - Fibroblast Growth Factor Basic Fragment 1-24 bovine

EMAIL THIS PAGE TO A FRIEND
F5895 Sigma-Aldrich

Fibroblast Growth Factor Basic Fragment 1-24 bovine

Synonym: FGF2

  • CAS Number 62031-54-3

  • Empirical Formula (Hill Notation) C118H173N31O33

  • Molecular Weight 2553.82

  •  MDL number MFCD00133317

Purchase

Properties

storage temp.   −20°C
SMILES string   CC(C)C[C@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCCN)NC(=O)[C@@H]1CCCN1C(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCCCN)NC(=O)[C@H](Cc2ccccc2)NC(=O)[C@H](Cc3c[nH]cn3)NC(=O)CNC(=O)[C@@H]4CCCN4C(=O)[C@@H]5CCCN5C(=O)[C@H](Cc6ccccc6)NC(=O)[C@H](C)NC(=O)CNC(=O)[C@H](CO)NC(=O)CNC(=O)CNC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@@H]7CCCN7C(=O)[C@H](CC(C)C)NC(=O)[C@H](C)NC(=O)[C@@H]8CCCN8)C(=O)N[C@@H](Cc9ccc(O)cc9)C(O)=O

Description

Application

Antibodies to FGF2 fragment 1-24 have been used for the immunohistochemical localization of FGF2 in various tissues.

Quality

HPLC shows two peaks which are believed to be different conformations of the same peptide.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
LK3641300

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?