• USA Home
  • F6022 - 3-Fluoro-3-deoxy-D-glucitol

EMAIL THIS PAGE TO A FRIEND
F6022 Sigma-Aldrich

3-Fluoro-3-deoxy-D-glucitol

  • CAS Number 34339-82-7

  • Empirical Formula (Hill Notation) C6H13FO5

  • Molecular Weight 184.16

Purchase

Properties

storage temp.   2-8°C

Description

Substrates

A substrate for NAD-linked sorbitol dehydrogenase.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?