• USA Home
  • F6037 - 2-Fluoro-2-deoxy-D-glucose 6-phosphate barium salt

EMAIL THIS PAGE TO A FRIEND
F6037 Sigma-Aldrich

2-Fluoro-2-deoxy-D-glucose 6-phosphate barium salt

Purchase

Properties

storage temp.   −20°C
SMILES string   OC(COP(O)(O)=O)C(O)C(O)C(F)C=O

Description

Biochem/physiol Actions

A metabolite of 2-fluoro-2-deoxy-D-glucose. Its formation is an indication of D-glucose transport, metabolism and steady state.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?