EMAIL THIS PAGE TO A FRIEND
F6426 Sigma-Aldrich

Foliosidine

≥96% (TLC), solid

  • CAS Number 2520-38-9

  • Empirical Formula (Hill Notation) C16H21NO5

  • Molecular Weight 307.34

  •  MDL number MFCD00274484

  •  PubChem Substance ID 24278438

Purchase

Properties

assay   ≥96% (TLC)
form   solid
storage temp.   2-8°C

Description

Biochem/physiol Actions

Anticonvulsant; causes hypothermia; prevents cardiac arrhythmia caused by electric stimulation of the atria or by aconitine.

Warning

Protect from light.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?