EMAIL THIS PAGE TO A FRIEND
F6768 Sigma-Aldrich

Fmoc-Gln(Trt)-OH

~95%

Synonym: Nα-Fmoc-Nδ-trityl-L-glutamine

  • CAS Number 132327-80-1

  • Empirical Formula (Hill Notation) C39H34N2O5

  • Molecular Weight 610.70

  •  Beilstein/REAXYS Number 4343953

  •  MDL number MFCD00077056

Purchase

Properties

InChI Key   WDGICUODAOGOMO-DHUJRADRSA-N
assay   ~95%
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

47674 Fmoc-Gln(Trt)-OH, ≥98.0% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?