• USA Home
  • F7013 - 4-Nitrobenzenediazonium tetrafluoroborate

EMAIL THIS PAGE TO A FRIEND
F7013 Sigma-Aldrich

4-Nitrobenzenediazonium tetrafluoroborate

Synonym: Azoic Diazo No. 37, Fast Red GG salt

  • CAS Number 456-27-9

  • Linear Formula C6H4N3O2 · BF4

  • Molecular Weight 236.92

  •  Colour Index Number 37035

  •  Beilstein/REAXYS Number 3599869

  •  EC Number 207-261-1

  •  MDL number MFCD00012005

Purchase

Properties

InChI Key   AFULQCYCQAHYIP-UHFFFAOYSA-N
composition   Dye content, >90%
mp   144-148 °C (dec.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3
RTECS 
CZ1723000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

294438 4-Nitrobenzenediazonium tetrafluoroborate, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?