• USA Home
  • F7383 - N-(3-[2-Furyl]acryloyl)-Gly-Leu amide

EMAIL THIS PAGE TO A FRIEND
F7383 Sigma-Aldrich

N-(3-[2-Furyl]acryloyl)-Gly-Leu amide

Synonym: FA-Gly-Leu-NH2

  • CAS Number 26400-33-9

  • Empirical Formula (Hill Notation) C15H21N3O4

  • Molecular Weight 307.34

  •  MDL number MFCD00077117

  •  PubChem Substance ID 24894940

Purchase

Properties

storage temp.   −20°C
SMILES string   CC(C)CC(NC(=O)CNC(=O)\C=C\c1ccco1)C(N)=O

Description

Substrates

Substrate for thermolysin.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?